Tisk…

Anti-ATP6V1C2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP17823C
ABGEAP17823CEA 10080 CZK
ABGEAP17823C
Anti-ATP6V1C2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ATP6V1C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu ATP6V1C2
Název antigenu ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2
Environmentally Preferable
Synonyma antigenu VMA5|ATP6C2
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 245973
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Průtoková cytometrie Yes
Epitop internal
Imunogen This ATP6V1C2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 276-304 amino acids from the Central region of human ATP6V1C2.
Počet aminokyselin 276 to 304
Sekvence DHKVKVTPLGNPDRC
Molekulární hmotnost 48.759 kDa
Čištění Protein A & Peptide affinity purification
Koncentrace 0.37
Skladovací pufr pH 7,4
Skladovací teplota Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR