Tisk…

Anti-TTK Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP17197A
ABGEAP17197AEA 10080 CZK
ABGEAP17197A
Anti-TTK Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR