Tisk…

Anti-PDE8A Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP1463C
ABGEAP1463CEA 10080 CZK
ABGEAP1463C
Anti-PDE8A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PDE8A (phosphodiesterase 8A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR