Tisk…

Anti-PDK3 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP14274B
ABGEAP14274BEA 10080 CZK
ABGEAP14274B
Anti-PDK3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PDK3 (pyruvate dehydrogenase kinase, isozyme 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR