Tisk…

Anti-PRSS8 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP14207C
ABGEAP14207CEA 10080 CZK
ABGEAP14207C
Anti-PRSS8 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PRSS8 (protease, serine, 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR