Tisk…

Anti-NR2F2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP14204A
ABGEAP14204AEA 10080 CZK
ABGEAP14204A
Anti-NR2F2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NR2F2 (nuclear receptor subfamily 2, group F, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR