Tisk…

Anti-CIDEA Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP14057B
ABGEAP14057BEA 10080 CZK
ABGEAP14057B
Anti-CIDEA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CIDEA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR