Tisk…

Anti-OR14C36 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP13761B
ABGEAP13761BEA 10080 CZK
ABGEAP13761B
Anti-OR14C36 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: OR14C36 (olfactory receptor, family 14, subfamily C, member 36)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR