Tisk…

Anti-RPS12 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP12967A
ABGEAP12967AEA 10080 CZK
ABGEAP12967A
Anti-RPS12 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RPS12 (ribosomal protein S12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR