Tisk…

Anti-SPNS3 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP12683B
ABGEAP12683BEA 10080 CZK
ABGEAP12683B
Anti-SPNS3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SPNS3 (spinster homolog 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR