Tisk…

Anti-CBLC Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP1257A
ABGEAP1257AEA 10080 CZK
ABGEAP1257A
Anti-CBLC Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CBLC (Cbl proto-oncogene C, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR