Tisk…

Anti-RHOJ Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP11763C
ABGEAP11763CEA 10080 CZK
ABGEAP11763C
Anti-RHOJ Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RHOJ (ras homolog family member J)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR