Tisk…

Anti-ATG16L2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP11695C
ABGEAP11695CEA 10080 CZK
ABGEAP11695C
Anti-ATG16L2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ATG16L2 (autophagy related 16-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR