Tisk…

Anti-SEC14L5 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP11616C
ABGEAP11616CEA 10080 CZK
ABGEAP11616C
Anti-SEC14L5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SEC14L5 (SEC14-like 5 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR