Tisk…

Anti-IL17F Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP10961A
ABGEAP10961AEA 10080 CZK
ABGEAP10961A
Anti-IL17F Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: IL17F (interleukin 17F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR