Tisk…

Anti-CLU Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP10943A
ABGEAP10943AEA 10080 CZK
ABGEAP10943A
Anti-CLU Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CLU (clusterin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR