Tisk…

Anti-UGT3A2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP10359B
ABGEAP10359BEA 10080 CZK
ABGEAP10359B
Anti-UGT3A2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: UGT3A2 (UDP glycosyltransferase 3 family, polypeptide A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR