Tisk…

Anti-NAA25 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP10270C
ABGEAP10270CEA 10080 CZK
ABGEAP10270C
Anti-NAA25 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NAA25 (N(alpha)-acetyltransferase 25, NatB auxiliary subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR