Tisk…

Anti-KDM6B Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP1022B
ABGEAP1022BEA 10080 CZK
ABGEAP1022B
Anti-KDM6B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KDM6B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR