Tisk…

Anti-CALCA Mouse Monoclonal Antibody

Dodavatel: Abgent
AM2021A
ABGEAM2021AEA 10080 CZK
ABGEAM2021A
Anti-CALCA Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CALCA (calcitonin-related polypeptide alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu CALCA
Environmentally Preferable
Název antigenu Calcitonin-related Polypeptide Alpha
Synonyma antigenu CT|Calcitonin-related Polypeptide alpha|CGRP|CALC1|KC|CGRP1|CGRP-I|Calcitonin
Druh protilátky Primární
Klonovalita Monoclonal
Spojení Unconjugated
Klon 406CT21.4.3
Reaktivita Human
Hostitel Mouse
Identifikační číslo genu 796
Izotyp IgM
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunogen This CALCA/CT antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 104-132 amino acids from human CALCA/CT.
Sekvence KRDMSSDLERDHRPC
Formulář Ascites
Molekulární hmotnost 15.466 kDa
Křížová adsorbce No
Skladovací pufr PBS with 0,09% (W/V) sodium azide, pH 7,4
Skladovací teplota Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR