Tisk…

trans-4-(Boc-amino)cyclohexanol

Dodavatel: abcr

TYB50390 TYB50390.0005
ABCRTYB50390EA 0 CZK
ABCRTYB50390 ABCRTYB50390.0005
trans-4-(Boc-amino)cyclohexanol
trans-4-(Boc-amino)cyclohexanol
Vzorec: C₁₁H₂₁NO₃
MDL Number: MFCD03844613
CAS číslo: 111300-06-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR