Tisk…

(±)-1-(1-Naphthyl)ethylamine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1-(Naphthalen-1-yl)ethanamine

L09342.06 L09342.14 AB178745.0025 AB178745.0005
L09342.06EA 1820 CZK
L09342.06 L09342.14 ABCRAB178745.0025 ABCRAB178745.0005
(±)-1-(1-Naphthyl)ethylamine 98%
(±)-1-(1-Naphthyl)ethylamine

Purity: 98% CAS No.: 42882-31-5 Molecular Formula: C12H13N Formula Weight: 171.24 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.6230 Boiling Point: 155-156°/15mm Beilstein Reference: 3197375 UN No.: 2922 MDL No.: MFCD00004014

Vzorec: C₁₂H₁₃N
Bod varu: 155…156 °C (15 mmHg)
Hustota: 1,063
Bod vzplanutí: >110 °C (230 °F)
MDL Number: MFCD00004014
CAS číslo: 42882-31-5
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR