Tisk…

(+)-Valencene

Dodavatel: abcr

AB168272
ABCRAB168272EA 0 CZK
ABCRAB168272
(+)-Valencene
(+)-Valencene
Vzorec: C₁₅H₂₄
MDL Number: MFCD00075884
CAS číslo: 4630-07-3
EINECS: 225-047-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR