Tisk…

8-Bromo-1-naphthoic acid

Dodavatel: abcr

AB145134.0001
ABCRAB145134.0001EA 0 CZK
ABCRAB145134.0001
8-Bromo-1-naphthoic acid
8-Bromo-1-naphthoic acid
Vzorec: C₁₁H₇BrO₂
MDL Number: MFCD00029357
CAS číslo: 1729-99-3
EINECS: 217-045-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR