Tisk…

2,3,4,4'-Tetrahydroxybenzophenone ≥98.0% (by titrimetric analysis)

Dodavatel: abcr
Varování

AB142192.0025
ABCRAB142192.0025EA 0 CZK
ABCRAB142192.0025
2,3,4,4'-Tetrahydroxybenzophenone ≥98.0% (by titrimetric analysis)
2,3,4,4'-Tetrahydroxybenzophenone
Vzorec: C₁₃H₁₀O₅
MDL Number: MFCD00083097
CAS číslo: 31127-54-5

Objednat

Výsledky specifikačního testu

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 218.0 to 222.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR