Tisk…

1-Bromo-2-naphthoic acid

Dodavatel: abcr

Synonyms: 1-Bromonaphthalene-2-carboxylic acid

AB138183.0001
ABCRAB138183.0001EA 0 CZK
ABCRAB138183.0001
1-Bromo-2-naphthoic acid
1-Bromo-2-naphthoic acid
Vzorec: C₁₁H₇BrO₂
MW: 251.0800 g/mol
MDL Number: MFCD00021408
CAS číslo: 20717-79-7
EINECS: 243-984-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR