Tisk…

3-Hydroxybenzonitrile ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol

B21702.06 B21702.14 AB132941.0005
B21702.06EA 1310 CZK
B21702.06 B21702.14 ABCRAB132941.0005
3-Hydroxybenzonitrile ≥99%
3-Hydroxybenzonitrile

Purity: 99% CAS No.: 873-62-1 Molecular Formula: C7H5NO Formula Weight: 119.12 Melting Point: 78-81° MDL No.: MFCD00002252

Vzorec: C₇H₅NO
Bod varu: 265 °C (1013 hPa)
Bod tání: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002252
CAS číslo: 873-62-1
EINECS: 212-845-4
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR