Tisk…

4-(Methylthio)benzyl alcohol 98

Dodavatel: Thermo Fisher Scientific

Synonyms: 4-(Methylthio)benzyl Alcohol

B22455.06 B22455.14 B22455.22 AB132566.0100
B22455.06EA 1730 CZK
B22455.06 B22455.14 B22455.22 ABCRAB132566.0100
4-(Methylthio)benzyl alcohol 98
4-(Methylthio)benzyl alcohol

Purity: 98% CAS No.: 3446-90-0 Molecular Formula: C8H10OS Formula Weight: 154.23 Flash Point: 110°(230°F) Melting Point: 41-43° Boiling Point: 108-109°/0.4mm MDL No.: MFCD00009706

Vzorec: C₈H₁₀OS
MW: 154.23 g/mol
Bod varu: 108…109 °C (0.4 mmHg)
Bod tání: 41…43 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009706
CAS číslo: 3446-90-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR