Tisk…

Diethyl aminomalonate hydrochloride 98%

Dodavatel: Thermo Fisher Scientific
Varování

A13681.09 A13681.18 A13681.30 AB115868.0250
A13681.09EA 1010 CZK
A13681.09 A13681.18 A13681.30 ABCRAB115868.0250
Diethyl aminomalonate hydrochloride 98%
Diethyl aminomalonate hydrochloride

Purity: 98% CAS No.: 13433-00-6 Molecular Formula: C7H13NO4.HCl Formula Weight: 211.65 Physical Form: Cryst powder Sensitivity: Hygroscopic Appearance: White to cream Melting Point: ca 165° dec. Beilstein Reference: 3568037 MDL No.: MFCD00012510

Vzorec: C₇H₁₄ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD00012510
CAS číslo: 13433-00-6
EINECS: 236-556-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR