Tisk…

Decamethylcyclopentasiloxane

Dodavatel: abcr

AB111012.2000 AB111012.0100
ABCRAB111012.2000EA 0 CZK
ABCRAB111012.2000 ABCRAB111012.0100
Decamethylcyclopentasiloxane
Decamethylcyclopentasiloxane
Vzorec: C₁₀H₃₀O₅Si₅
Bod varu: 90 °C (13 hPa)
Bod tání: –44 °C
Hustota: 0,958 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00046966
CAS číslo: 541-02-6
EINECS: 208-764-9
Merck Index: 12,02905

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR