Tisk…

Anti-SRRT Mouse Polyclonal Antibody

Dodavatel: Abcam
AB88392-50
ABCAAB88392-50EA 28530 CZK
ABCAAB88392-50
Anti-SRRT Mouse Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SRRT (serrate RNA effector molecule homolog (Arabidopsis))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR