Tisk…
Anti-CD48 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-102]
Dodavatel: Abcam
AB76904-50
ABCAAB76904-50EA 10870 CZK
ABCAAB76904-50
Anti-CD48 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-102]
Protilátky
Upozornení:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production
More Product Information
Objednat
Produktová specifikace
Symbol antigenu CD48
Environmentally Preferable
Název antigenu CD48 molecule
Druh protilátky Primární
Klonovalita Monoclonal
Spojení APC (Allophycocyanin)
Klon MEM-102
Reaktivita Human
Hostitel Mouse
Izotyp IgG1
Western blot Yes
Imunochemie Yes
Imunoprecipitace Yes
Průtoková cytometrie Yes
Skladovací teplota Cool
Transportní teplota Ice