Tisk…
Anti-CFLAR Rabbit polyclonal antibody unconjugated
Dodavatel: Abcam
AB6144-100
ABCAAB6144-100EA 19950 CZK
ABCAAB6144-100
Anti-CFLAR Rabbit polyclonal antibody unconjugated
Protilátky
Upozornení:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production
More Product Information
Objednat
Produktová specifikace
Symbol antigenu CFLAR
Environmentally Preferable
Název antigenu CASP8 and FADD-like apoptosis regulator
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Skladovací teplota Cool
Transportní teplota Ice