Tisk…
Anti-Leptin Rabbit Polyclonal Antibody
Dodavatel: Abcam
AB3583-50
ABCAAB3583-50EA 20490 CZK
ABCAAB3583-50
Anti-Leptin Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal to Leptin.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any ofMore Product Information
Objednat
Produktová specifikace
Symbol antigenu LEP
Název antigenu leptin
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Skladovací teplota Cool
Transportní teplota Ice