Tisk…

Dihydrotestosterone (DHT) ELISA Kit

Dodavatel: Abcam

AB287824-96
ABCAAB287824-96EA 26900 CZK
ABCAAB287824-96
Dihydrotestosterone (DHT) ELISA Kit
Test ELISY

A competitive ELISA kit for the quantitative measurement of dihydrotestosterone in biological fluids.


  • Detection range: 39,063 to 2500 pg/ml
  • Sensitivity: <23,438 pg/ml
  • Assay precision: Intra-assay: CV <8%; Inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed


Easy, convenient and time-saving method to measure the level of Dihydrotestosterone in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.


Dihydrotestosterone (DHT), also known as androstanolone or stanolone, is an endogenous androgen sex steroid and hormone. DHT is biologically important for sexual differentiation of the male genitalia during embryogenesis, maturation of the penis and scrotum at puberty, growth of facial, body, and pubic hair, and development and maintenance of the prostate gland and seminal vesicles.


The enzyme 5α-reductase catalyses the formation of DHT from testosterone by mediates the reduction of the C4-5 double bond of testosterone. Relative to testosterone, DHT is considerably more potent as an agonist of the androgen receptor (AR).


Dihydrotestosterone ELISA kit is a competitive ELISA assay for the quantitative measurement of dihydrotestosterone in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of dihydrotestosterone captured from the samples.


Upozornení:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.


Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR