Tisk…
Anti-RANGAP1 Goat Polyclonal Antibody
Dodavatel: Abcam
AB2081-100
ABCAAB2081-100EA 19600 CZK
ABCAAB2081-100
Anti-RANGAP1 Goat Polyclonal Antibody
Protilátky
Upozornení:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production
More Product Information
Objednat
Produktová specifikace
Symbol antigenu RANGAP1
Environmentally Preferable
Název antigenu Ran GTPase activating protein 1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Mouse
Hostitel Goat
Izotyp IgG
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Skladovací teplota Cool
Transportní teplota Ice