Tisk…

Anti-LGALS1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
AB112525-100
ABCAAB112525-100EA 19600 CZK
ABCAAB112525-100
Anti-LGALS1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LGALS1 (lectin, galactoside-binding, soluble, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR