Tisk…

Anti-Spectrin beta 5 Rabbit Polyclonal Antibody

Dodavatel: Abcam
AB111223-200
ABCAAB111223-200EA 18890 CZK
ABCAAB111223-200
Anti-Spectrin beta 5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Spectrin beta 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR