Tisk…

Anti-TSPO Rabbit Monoclonal Antibody [clone: EPR5384]

Dodavatel: Abcam
AB109497-100
ABCAAB109497-100EA 23520 CZK
ABCAAB109497-100
Anti-TSPO Rabbit Monoclonal Antibody [clone: EPR5384]
Protilátky
Type: Primary
Antigen: TSPO (translocator protein (18kDa))
Clonality: monoclonal
Clone: EPR5384
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR