Tisk…

Anti-IGFBP2 Rabbit Monoclonal Antibody [clone: EPR3380]

Dodavatel: Abcam
AB109284-100
ABCAAB109284-100EA 29020 CZK
ABCAAB109284-100
Anti-IGFBP2 Rabbit Monoclonal Antibody [clone: EPR3380]
Protilátky
Type: Primary
Antigen: IGFBP2 (insulin-like growth factor binding protein 2, 36kDa)
Clonality: monoclonal
Clone: EPR3380
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR