Tisk…
XFD532 NHS Ester [Same Structure to Alexa Fluor™ 532 NHS ester]
Dodavatel: AAT BIOQUEST
71904
AATB71904EA 29350 CZK
AATB71904
XFD532 NHS Ester [Same Structure to Alexa Fluor™ 532 NHS ester]
XFD532 NHS Ester
XFD532 NHS Ester (Succinimidyl Ester) is the same molecule to the Alexa Fluor® 532 NHS Ester (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Upozornení: For Research Use Only