Tisk…
Amplite® Tyramide (TSA) Signal Booster
Dodavatel: AAT BIOQUEST
44502
AATB44502EA 3550 CZK
AATB44502
Amplite® Tyramide (TSA) Signal Booster
Činidla pro nukleové kyseliny Nucleic Acid Modification and Labelling
Tyramide signal amplification (TSA) is a technique used to enhance the signal of target molecules, such as antibodies or nucleic acid probes, in immunohistochemistry and in situ hybridisation.
  • Stable for 12 months
Quality tested in-house.
Upozornení: For research use only.