Tisk…

N-FMOC-ethylenediamine

Dodavatel: AAT BIOQUEST
Varování

4016 4017
AATB4016EA 20520 CZK
AATB4016 AATB4017
N-FMOC-ethylenediamine
N-FMOC-ethylenediamine

N-FMOC-ethylenediamine is a monoprotected diamine that is widely used as a building block in bioconjugate chemistry.


  • Excellent building block
  • Multiple applications

Vzorec: C₁₇H₁₈N₂O₂
CAS číslo: 166410-32-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b09e6205cb9cebfdbad824036dc671fda953c5cc&vid=0c52a0ae9adcb951ee59e7ed1e929c41c49925b3&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR