Tisk…
Rapamycin-LC-Fluorescein
Dodavatel: AAT BIOQUEST
36706
AATB36706EA 10730 CZK
AATB36706
Rapamycin-LC-Fluorescein
Rapamycin-LC-Fluorescein
Rapamycin-LC-Fluorescein is an excellent fluorescent probe to characterize FK506-binding proteins by fluorescence polarization.
  • Stable for 24 months
Quality tested in-house
Upozornení: For Research Use Only