Tisk…

Thioflavin T ≥95%

Dodavatel: AAT BIOQUEST
Varování

Synonyms: CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T

23060
AATB23060EA 3210 CZK
AATB23060
Thioflavin T ≥95%
Thioflavin T

Thioflavin T (ThT), also called Basic Yellow 1 or CI 49005, is a benzothiazole salt.

Vzorec: C₁₇H₁₉ClN₂S
MW: 318,86 g/mol
MDL Number: MFCD00011944
CAS číslo: 2390-54-7
ADR: 5,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR