Tisk…
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Dodavatel: AAT BIOQUEST
22326
AATB22326EA 18970 CZK
AATB22326
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Test Buněčná stanovení
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
  • FITC filter set compatible
  • Bright fluorescence
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit uses XdU which is incorporated into cellular DNA during DNA synthesis. After fixing cells, the incorporated XdU is labelled with iFluor® 488 MTA. The resulted iFluor® 488-labeled DNA formed in cells is imaged with a FITC filter set. Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay..
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.