Tisk…

Dextran, M.W. 20,000 TRITC-labeled

Dodavatel: AAT BIOQUEST

21706
AATB21706EA 7790 CZK
AATB21706
Dextran, M.W. 20,000 TRITC-labeled
Dextran

TRITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.


  • Well-retained in cells
  • 20,000 mw
  • Multiple applications


From Eukaryotic cells


Upozornení: For research use only.

Vzorec: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS číslo: 9004-54-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR