Tisk…

Nicotinic acid adenine dinucleotide phosphate tetrasodium salt 95

Dodavatel: AAT BIOQUEST
Varování

20999
AATB20999EA 4060 CZK
AATB20999
Nicotinic acid adenine dinucleotide phosphate tetrasodium salt 95
Nicotinic acid adenine dinucleotide phosphate tetrasodium salt

NAADP is a calcium mobilizing agent.

Vzorec: C₂₁H₂₃N₆Na₄O₁₈P₃
MDL Number: MFCD00274064
CAS číslo: 5502-96-5
ADR: 5 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR