Tisk…
Annexin V-iFluor® 594 Conjugate
Dodavatel: AAT BIOQUEST
20073
AATB20073EA 8190 CZK
AATB20073
Annexin V-iFluor® 594 Conjugate
Test Buněčná stanovení
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
  • High stability
  • In-house quality control
  • Multiple research applications
Upozornení: For research use only.