Tisk…

4-[(Dipropylamino)sulphonyl]benzoic acid sodium salt, water soluble, ReadiUse™

Dodavatel: AAT BIOQUEST
Varování

20061
AATB20061EA 6140 CZK
AATB20061
4-[(Dipropylamino)sulphonyl]benzoic acid sodium salt, water soluble, ReadiUse™
4-[(Dipropylamino)sulphonyl]benzoic acid sodium salt

Probenecid is an inhibitor of organic-anion transporters located in cell membranes.


  • No naoh required
  • Ready to use in three formats
  • Decreases calcium assay background


Upozornení: For research use only.

Vzorec: C₁₃H₁₈NO₄SNa
CAS číslo: 23795-03-1
ADR: 5,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR